3-Bromo-4-methoxybenzyl alcohol 98%

Supplier: Ambeed
Warning

A157377-10G A157377-250MG A157377-25G A157377-1G
103804-998EA 12 USD
103804-998 103843-200 77273-170 77273-172 77300-256 77319-018
3-Bromo-4-methoxybenzyl alcohol 98%
3-Bromo-4-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
Boiling Pt: 307.3 °C (760 mm Hg)
Melting Pt: 59…63 °C
Flash Pt: 139.646 °C
Storage Temperature: Ambient
MDL Number: MFCD01861398
CAS Number: 38493-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR