3-Bromo-2-methylbenzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2-methyl-benzenemethanol, (3-Bromo-o-tolyl)methanol, (3-Bromo-2-methylphenyl)methanol

A266328-1G A266328-25G A266328-5G A266328-100G A266328-10G A266328-250MG
103812-796EA 8 USD
103812-796 103852-698 77092-400 77092-402 77092-404 77166-328 77166-330 77166-332
3-Bromo-2-methylbenzyl alcohol 97%
3-Bromo-2-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MW: 201.06 g/mol
Melting Pt: 101…104 °C
Storage Temperature: Ambient
MDL Number: MFCD11847531
CAS Number: 83647-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR