3-Bromo-2,5-difluorobenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (3-Bromo-2,5-difluorophenyl)methanol

A363540-250MG A363540-1G
103826-348EA 81 USD
103826-348 103837-366 77015-900 77306-750
3-Bromo-2,5-difluorobenzyl alcohol 98%
3-Bromo-2,5-difluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅BrF₂O
MW: 223.015 g/mol
Boiling Pt: 254.0±35.0 °C (760 mm Hg)
Flash Pt: 107.4 °C
Storage Temperature: Ambient
MDL Number: MFCD27923603
CAS Number: 1159186-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR