3-(Boc-amino)phenylboronic acid 97%

Supplier: Ambeed
Warning

A118028-100G A118028-10G A118028-250MG A118028-25G A118028-1G A118028-5G
103798-976EA 8 USD
103798-976 103824-870 76797-204 76797-206 76797-208 76797-210 77311-654 77311-656
3-(Boc-amino)phenylboronic acid 97%
3-(Boc-amino)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₄
Melting Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD03411945
CAS Number: 380430-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR