1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose 97%

Supplier: Ambeed
Warning

A225244-100G A225244-500G A225244-25G A225244-5G A225244-10G
103809-594EA 196 USD
103809-594 103850-060 76722-090 76722-092 77303-734 77303-736 77879-365
1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose 97%
1,2:5,6-Di-O-isopropylidene-alpha-D-glucofuranose

Precaución: for Research Use Only

Formula: C₁₂H₂₀O₆
Boiling Pt: 362.8 °C (760 mm Hg)
Melting Pt: 107…113 °C
Flash Pt: 173.2 °C
Storage Temperature: Freezer
MDL Number: MFCD00005544
CAS Number: 582-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR