(-)-Corey lactone 98%

Supplier: Ambeed
Warning

Synonyms: (-)-6β-Hydroxymethyl-7α-hydroxy-cis-2-oxabicyclo[3.3.0]octan-3-one, (3aR,4S,5R,6aS)-5-Hydroxy-4-(hydroxymethyl)hexahydro-2H-cyclopenta[b]furan-2-one, (3aR,4S,5R,6aS)-(-)-Hexahydro-5-hydroxy-4-(hydroxymethyl)-2H-cyclopenta[b]furan-2-one, (-)-Corey lactone diol

A110303-10G A110303-250MG A110303-25G A110303-1G A110303-5G
103797-648EA 13 USD
103797-648 103823-456 77135-988 77135-990 77135-992 77310-254 77310-256
(-)-Corey lactone 98%
(-)-Corey lactone

Precaución: for Research Use Only

Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Boiling Pt: 406.6 °C (760 mm Hg)
Melting Pt: 117…119 °C
Flash Pt: 172.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00075234
CAS Number: 32233-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR