(-)-Corey lactone-4-phenylbenzoate 98%

Supplier: Ambeed
Warning

Synonyms: (3aR,4S,5R,6aS)-4-(Hydroxymethyl)-2-oxohexahydro-2H-cyclopenta[b]furan-5-yl 4-Phenylbenzoate, (3aR,4S,5R,6aS)-Hexahydro-4-(hydroxymethyl)-5-(4-phenylbenzoyloxy)-2H-cyclopenta[b]furan-2-one, (3aR,4S,5R,6aS)-4-(Hydroxymethyl)-2-oxohexahydro-2H-cyclopenta[b]furan-5-yl Biphenyl-4-carboxylate

A454000-1G A454000-5G
103827-818EA 30 USD
103827-818 103838-888 76842-724 76842-726
(-)-Corey lactone-4-phenylbenzoate 98%
(-)-Corey lactone-4-phenylbenzoate

Precaución: for Research Use Only

Formula: C₂₁H₂₀O₅
MW: 352.39 g/mol
Boiling Pt: 584.8 °C (760 mm Hg)
Melting Pt: 128…134 °C at N/A mmHg
Flash Pt: 210.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00078077
CAS Number: 31752-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR