3-Aminophenylboronic acid hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (3-Aminophenyl)boronic acid hydrochloride, 3-Aminobenzeneboronic acid hydrochloride

A108286-1G A108286-5G
103797-322EA 10 USD
103797-322 103823-102 77309-944 77309-946
3-Aminophenylboronic acid hydrochloride 97%
3-Aminophenylboronic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₉BClNO₂
MW: 173.41 g/mol
Melting Pt: >320 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191748
CAS Number: 85006-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR