(m-Acrylamidophenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Acrylamidophenylboronic acid, (3-Acrylamidophenyl)boronic acid

A299413-1G A299413-5G A299413-10G A299413-250MG
103814-228EA 13 USD
103814-228 103835-374 76838-066 76838-068 77154-154 77154-156
(m-Acrylamidophenyl)boronic acid 98%
(m-Acrylamidophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BNO₃
Melting Pt: 129…146 °C
Storage Temperature: Ambient
MDL Number: MFCD09025755
CAS Number: 99349-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR