3,5-Di-tert-butylphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Di-tert-butylbenzeneboronic acid, 3,5-bis(tert-butyl)phenylboronic acid, (3,5-Di-tert-butylphenyl)boronic acid

A226087-100G A226087-10G A226087-1G A226087-25G A226087-5G A226087-500G
103819-220EA 11 USD
103819-220 103850-122 76874-116 76874-118 77088-842 77088-844 77303-758 77883-587
3,5-Di-tert-butylphenylboronic acid 98%
3,5-Di-tert-butylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₃BO₂
Melting Pt: 222…226 °C
Flash Pt: 155.229 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07780212
CAS Number: 197223-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR