3,5-Dibromobenzyl alcohol 98%

Supplier: Ambeed
Warning

A179799-1G A179799-100MG A179799-10G A179799-250MG A179799-25G A179799-5G A179799-100G
103807-468EA 15 USD
103807-468 103845-820 77083-296 77138-666 77138-668 77138-670 77301-634 77301-636 77685-405
3,5-Dibromobenzyl alcohol 98%
3,5-Dibromobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆Br₂O
MW: 265.93 g/mol
Boiling Pt: 327.9 °C (760 mm Hg)
Melting Pt: 107…108 °C
Flash Pt: 152 °C
Storage Temperature: Ambient
MDL Number: MFCD01632143
CAS Number: 145691-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR