3,5-Bis(trifluoromethyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Bis(trifluoromethyl)benzeneboronic acid, (3,5-Bis(trifluoromethyl)phenyl)metaboric acid, (3,5-Bis(trifluoromethyl)phenyl)boronic acid, Dihydroxy(3,5-bis(trifluoromethyl)phenyl)borane

A200674-100G A200674-10G A200674-1G A200674-25G A200674-5G
103847-866EA 13 USD
103847-866 77192-280 77192-282 77192-284 77302-710 77302-712
3,5-Bis(trifluoromethyl)phenylboronic acid 98%
3,5-Bis(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₅BF₆O₂
Flash Pt: 64.281 °C
Storage Temperature: Ambient
MDL Number: MFCD00051850
CAS Number: 73852-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR