3,4-Dimethylphenylhydrazinium chloride 97%

Supplier: Ambeed
Warning

Synonyms: (3,4-Dimethylphenyl)hydrazine hydrochloride, 3,4-Dimethylphenylhydrazine hydrochloride, 1-(3,4-Dimethylphenyl)hydrazine hydrochloride

A149620-100G A149620-10G A149620-25G A149620-5G
103842-182EA 6 USD
103842-182 77190-748 77190-750 77299-738 77299-740
3,4-Dimethylphenylhydrazinium chloride 97%
3,4-Dimethylphenylhydrazinium chloride

Precaución: for Research Use Only

Formula: C₈H₁₃ClN₂
MW: 172.66 g/mol
Melting Pt: 195…200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052270
CAS Number: 60481-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR