3,4-Difluorophenylboronic acid 98%

Supplier: Ambeed
Warning

A314119-25G A314119-100G A314119-10G A314119-500G A314119-5G
103835-942EA 7 USD
103835-942 76838-752 77099-742 77099-744 77099-746 77306-108
3,4-Difluorophenylboronic acid 98%
3,4-Difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BF₂O₂
Boiling Pt: 265.4 °C (760 mm Hg)
Melting Pt: 289…290 °C
Storage Temperature: Ambient
MDL Number: MFCD00807405
CAS Number: 168267-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR