3,4,5-Trifluorophenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(3,4,5-Trifluorophenyl)metaboric acid, Dihydroxy(3,4,5-trifluorophenyl)borane, 3,4,5-Trifluorobenzeneboronic acid, (3,4,5-Trifluorophenyl)boronic acid
A171057-25G
A171057-5G
A171057-100G
A171057-10G
A171057-1G
A171057-500G
103806-584EA
5
USD
103806-584
103844-858
77081-978
77081-980
77096-508
77096-510
77301-094
77550-102
3,4,5-Trifluorophenylboronic acid 98%
3,4,5-Trifluorophenylboronic acid
Precaución: for Research Use Only
Formula:
C₆H₄BF₃O₂ MW: 175.9 g/mol Melting Pt: 290…295 °C Flash Pt: 113.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093069 CAS Number: 143418-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...