3,4,5-Trichlorophenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (3,4,5-Trichlorophenyl)boronic acid

A223655-5G A223655-10G A223655-250MG A223655-1G
103819-050EA 10 USD
103819-050 103849-930 77088-554 77112-796 77112-798 77303-656
3,4,5-Trichlorophenylboronic acid 95%
3,4,5-Trichlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BCl₃O₂
MW: 225.27 g/mol
Boiling Pt: 379.1 °C (760 mm Hg)
Flash Pt: 183.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095126
CAS Number: 862248-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR