3-(Trimethylsilyl)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (3-(Trimethylsilyl)phenyl)boronic acid

A113773-250MG A113773-25G A113773-5G
103798-290EA 15 USD
103798-290 103824-134 77245-840 77245-842 77310-950
3-(Trimethylsilyl)phenylboronic acid 97%
3-(Trimethylsilyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₅BO₂Si
Boiling Pt: 290.2 °C (760 mm Hg)
Melting Pt: 145…150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03093884
CAS Number: 177171-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR