3-(Trifluoromethyl)benzyl alcohol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(3-(Trifluoromethyl)phenyl)methanol, 3-(Hydroxymethyl)benzotrifluoride
A224961-100G
A224961-10G
A224961-500G
A224961-25G
A224961-5G
103850-040EA
7
USD
103850-040
76975-958
76975-960
76975-962
77088-706
77088-708
3-(Trifluoromethyl)benzyl alcohol 98%
3-(Trifluoromethyl)benzyl alcohol
Precaución: for Research Use Only
Formula:
C₈H₇F₃O MW: 176.14 g/mol Boiling Pt: 214.4 °C (760 mm Hg) Flash Pt: 84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004645 CAS Number: 349-75-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...