3-(Trifluoromethyl)phenylhydrazinium chloride 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Trifluoromethyl)phenylhydrazine hydrochloride, 3-Hydrazinobenzotrifluoride hydrochloride, (3-(Trifluoromethyl)phenyl)hydrazine hydrochloride, 1-(3-(Trifluoromethyl)phenyl)hydrazine hydrochloride

A157101-100G A157101-10G A157101-25G A157101-1G A157101-5G
103804-962EA 5 USD
103804-962 103843-162 77138-002 77138-004 77300-232 77318-978 77318-980
3-(Trifluoromethyl)phenylhydrazinium chloride 97%
3-(Trifluoromethyl)phenylhydrazinium chloride

Precaución: for Research Use Only

Formula: C₇H₈ClF₃N₂
MW: 212.6 g/mol
Flash Pt: 107.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00100503
CAS Number: 3107-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR