(3-(Pyridin-3-yl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A230542-10G A230542-250MG A230542-1G
103819-554EA 23 USD
103819-554 103850-454 77013-492 77013-494 77089-314
(3-(Pyridin-3-yl)phenyl)boronic acid 98%
(3-(Pyridin-3-yl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀BNO₂
Boiling Pt: 439 °C (760 mm Hg)
Melting Pt: 178…181 °C
Flash Pt: 220 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16658918
CAS Number: 351422-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR