(3-(Diphenylamino)phenyl)boronic acid 95%

Supplier: Ambeed
Warning

A648768-1G A648768-5G A648768-250MG A648768-25G
103840-390EA 162 USD
103840-390 76844-550 76844-552 77018-334 77891-286
(3-(Diphenylamino)phenyl)boronic acid 95%
(3-(Diphenylamino)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₈H₁₆BNO₂
Boiling Pt: 494.4 °C at N/A mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD22570743
CAS Number: 943899-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR