3-(Difluoromethyl)benzeneboronic acid 98%

Supplier: Ambeed
Danger

Synonyms: (3-(Difluoromethyl)phenyl)boronic acid

A173485-100MG A173485-250MG A173485-1G A173485-5G
103806-820EA 56 USD
103806-820 103845-120 76813-290 76813-292 77082-362 77301-274
3-(Difluoromethyl)benzeneboronic acid 98%
3-(Difluoromethyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₂
Boiling Pt: 312 °C (760 mm Hg)
Melting Pt: 38…40 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09864670
CAS Number: 854690-87-2
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR