[3-(Bromomethyl)oxetan-3-yl]methanol 90%

Supplier: Ambeed
Warning

Synonyms: (3-(Bromomethyl)oxetan-3-yl)methanol

A314658-1G A314658-25G A314658-5G A314658-100G A314658-10G
103825-042EA 8 USD
103825-042 103835-964 76838-780 76838-782 76838-784 77277-602 77277-604
[3-(Bromomethyl)oxetan-3-yl]methanol 90%
[3-(Bromomethyl)oxetan-3-yl]methanol

Precaución: for Research Use Only

Formula: C₅H₉BrO₂
MW: 181.04 g/mol
Boiling Pt: 242.4 °C (760 mm Hg)
Flash Pt: 100.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09800598
CAS Number: 22633-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR