(3-(Benzyloxy)isoxazol-5-yl)methanol 98%

Supplier: Ambeed
Warning

A139040-100MG A139040-250MG A139040-1G
103802-690EA 79 USD
103802-690 76823-348 76823-350 77315-764
(3-(Benzyloxy)isoxazol-5-yl)methanol 98%
(3-(Benzyloxy)isoxazol-5-yl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₃
MW: 205.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23135734
CAS Number: 123320-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR