(3-(4-Bromophenyl)isoxazol-5-yl)methanol 97%

Supplier: Ambeed
Warning

A395820-1G A395820-5G
103826-966EA 80 USD
103826-966 103838-004 76841-422 76841-424
(3-(4-Bromophenyl)isoxazol-5-yl)methanol 97%
(3-(4-Bromophenyl)isoxazol-5-yl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO₂
MW: 254.08 g/mol
Boiling Pt: 410.4±35.0 °C (760 mm Hg)
Flash Pt: 202 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06656791
CAS Number: 206055-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR