3-(Hydroxymethyl)phenylboronic acid pinacol ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-[3-(Hydroxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Hydroxymethylphenylboronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl alcohol, (3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol, 3-(Hydroxymethyl)phenylboronic acid Pinacol ester, 3-(Hydroxymethyl)benzeneboronic acid pinacol ester
A117943-100G
A117943-250MG
A117943-25G
A117943-1G
A117943-10G
103798-958EA
12
USD
103798-958
103824-848
76822-230
76822-232
76822-234
77311-630
77683-060
3-(Hydroxymethyl)phenylboronic acid pinacol ester 97%
3-(Hydroxymethyl)phenylboronic acid pinacol ester
Precaución: For research use only.
Formula:
C₁₃H₁₉BO₃ MW: 234.10 g/mol Boiling Pt: 363.7 °C (760 mm Hg) Melting Pt: 42…46 °C Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09266196 CAS Number: 443776-76-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...