3-(2-Methoxy-2-oxoethyl)phenylboronic acid 97%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: (3-(2-Methoxy-2-oxoethyl)phenyl)boronic acid

A192567-100MG A192567-250MG
103816-458EA 115 USD
103816-458 103847-066 77012-400 77683-511
3-(2-Methoxy-2-oxoethyl)phenylboronic acid 97%, Ambeed.Inc
3-(2-Methoxy-2-oxoethyl)phenylboronic acid

Precaución: For research use only.

Formula: C₉H₁₁BO₄
Boiling Pt: 358.3 °C (760 mm Hg)
Melting Pt: 108…111 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03425946
CAS Number: 643094-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR