3-(2-Hydroxyethyl)benzeneboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (3-(2-Hydroxyethyl)phenyl)boronic acid

A142202-250MG A142202-1G
103803-120EA 78 USD
103803-120 77272-520 77316-336
3-(2-Hydroxyethyl)benzeneboronic acid 97%
3-(2-Hydroxyethyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
MW: 165.98 g/mol
Boiling Pt: 376 °C (760 mm Hg)
Flash Pt: 181.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095123
CAS Number: 647853-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR