Sulbactam 98%

Supplier: Ambeed
Warning

A637045-1G A637045-250MG A637045-25G
103840-318EA 52 USD
103840-318 77602-610 77613-980 77613-982
Sulbactam 98%
Sulbactam

Precaución: Sealed in dry, store in freezer, under –20 °C.

Formula: C₈H₁₁NO₅S
Boiling Pt: 356 °C (760 mm Hg)
Melting Pt: 154…157 °C
Flash Pt: 297.1 °C
Storage Temperature: Freezer
MDL Number: MFCD00867005
CAS Number: 68373-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR