(2S,5S)-tert-Butyl-2,5-dimethylpiperazine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: (2S,5S)-tert-Butyl 2,5-dimethylpiperazine-1-carboxylate

A103532-100MG A103532-10G A103532-250MG A103532-1G A103532-5G
103796-570EA 51 USD
103796-570 103822-276 76821-340 76821-342 76821-344 77309-150 77309-152
(2S,5S)-tert-Butyl-2,5-dimethylpiperazine-1-carboxylate 97%
(2S,5S)-tert-Butyl-2,5-dimethylpiperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08686668
CAS Number: 1238951-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR