(2S,5S)-1-(tert-Butoxycarbonyl)-5-methylpyrrolidine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A216963-1G A216963-5G A216963-10G A216963-250MG A216963-25G
103818-590EA 22 USD
103818-590 103849-316 77087-928 77087-930 77139-774 77139-776 77303-440
(2S,5S)-1-(tert-Butoxycarbonyl)-5-methylpyrrolidine-2-carboxylic acid 97%
(2S,5S)-1-(tert-Butoxycarbonyl)-5-methylpyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.27 g/mol
Boiling Pt: 343.2±35.0 °C (760 mm Hg)
Flash Pt: 161.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13185989
CAS Number: 334769-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR