(2S,5R)-tert-Butyl 2,5-dimethylpiperazine-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2S,5R)-1-Boc-2,5-dimethylpiperazine, (2S,5R)-2,5-Dimethylpiperazine, N1-BOC protected
A539683-10G
A539683-250MG
A539683-1G
A539683-25G
A539683-5G
A539683-100G
103828-622EA
14
USD
103828-622
103839-744
76830-490
76830-492
76843-792
76843-794
76843-796
77569-054
(2S,5R)-tert-Butyl 2,5-dimethylpiperazine-1-carboxylate 97%
(2S,5R)-tert-Butyl 2,5-dimethylpiperazine-1-carboxylate
Precaución: For research use only.
Formula:
C₁₁H₂₂N₂O₂ MW: 214.30 g/mol Boiling Pt: 280 °C (760 mm Hg) Flash Pt: 123 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08686667 CAS Number: 548762-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...