(2S,5R)-5-((tert-Butoxycarbonyl)amino)tetrahydro-2H-pyran-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A179604-100MG A179604-250MG A179604-1G
103807-444EA 121 USD
103807-444 77011-946 77011-948 77083-264
(2S,5R)-5-((tert-Butoxycarbonyl)amino)tetrahydro-2H-pyran-2-carboxylic acid 95%
(2S,5R)-5-((tert-Butoxycarbonyl)amino)tetrahydro-2H-pyran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₅
MW: 245.27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20261452
CAS Number: 603130-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR