(2S,4S)-4-Fluoropyrrolidine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A114649-100MG A114649-10G A114649-250MG A114649-5G A114649-1G
103798-440EA 55 USD
103798-440 103824-284 77160-910 77160-912 77160-914 77297-318 77311-104
(2S,4S)-4-Fluoropyrrolidine-2-carboxylic acid 97%
(2S,4S)-4-Fluoropyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₈FNO₂
Boiling Pt: 265.1 °C (760 mm Hg)
Flash Pt: 114.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683983
CAS Number: 2438-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR