N-t-BOC-cis-4-Fluoro-L-proline methyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Boc-cis-4-F-Pro-OMe, (2S,4S)-1-tert-Butyl 2-methyl 4-fluoropyrrolidine-1,2-dicarboxylate

A103775-250MG A103775-1G A103775-5G A103775-25G
103796-610EA 14 USD
103796-610 103822-320 77025-314 77309-186 77309-188 77871-395
N-t-BOC-cis-4-Fluoro-L-proline methyl ester 95%
N-t-BOC-cis-4-Fluoro-L-proline methyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₈FNO₄
Boiling Pt: 296.525 °C (760 mm Hg)
Flash Pt: 133.134 °C
Storage Temperature: Ambient
MDL Number: MFCD11054130
CAS Number: 203866-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR