N-Boc-cis-4-Methyl-L-Proline 97%

Supplier: Ambeed
Warning

Synonyms: (4S)-1-(tert-Butoxycarbonyl)-4-methyl-L-proline, (2S,4S)-1-(tert-Butoxycarbonyl)-4-methylpyrrolidine-2-carboxylic acid

A186163-100MG A186163-10G A186163-250MG
103815-922EA 98 USD
103815-922 103846-496 77138-902 77138-904 77138-906
N-Boc-cis-4-Methyl-L-Proline 97%
N-Boc-cis-4-Methyl-L-Proline

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.273 g/mol
Boiling Pt: 343 °C (760 mm Hg)
Flash Pt: 161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07784043
CAS Number: 364750-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR