(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-Cis-4-F-Pro-OH, N-Boc-cis-4-fluoro-L-proline, N-(tert-Butoxycarbonyl)-cis-4-fluoro-L-proline, (2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoropyrrolidine-2-carboxylic acid, (2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid, (2S,4S)-1-Boc-4-fluoro-2-pyrrolidinecarboxylic Acid, (2S,4S)-1-[(tert-butoxy)carbonyl]-4-fluoropyrrolidine-2-carboxylic acid
A112064-100G
A112064-10G
A112064-250MG
A112064-25G
A112064-1G
A112064-5G
103797-946EA
8
USD
103797-946
103823-778
77008-964
77008-966
77008-968
77008-970
77310-584
77310-586
(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid 97%
(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₆FNO₄ MW: 233.24 g/mol Boiling Pt: 346 °C (760 mm Hg) Melting Pt: 157…161 °C Flash Pt: 163 °C Storage Temperature: Ambient |
MDL Number:
MFCD04973957 CAS Number: 203866-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...