(2S,4S)-1-(Benzyloxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2S,4S)-1-((Benzyloxy)carbonyl)-4-hydroxypyrrolidine-2-carboxylic acid, Z-cis-Hyp-OH, Cbz-cis-Hyp-OH

A102685-100MG A102685-250MG A102685-1G A102685-5G
103796-408EA 28 USD
103796-408 103822-106 76780-568 76780-570 77308-974 77308-976
(2S,4S)-1-(Benzyloxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid 98%
(2S,4S)-1-(Benzyloxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₅
MW: 265.265 g/mol
Boiling Pt: 486.9 °C (760 mm Hg)
Flash Pt: 248.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00238423
CAS Number: 13504-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR