N-Boc-trans-4-(fmoc-amino)-L-proline 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-BOC-trans-4-N-FMOC-amino-L-proline, (2S,4R)-4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-1-(tert-butoxycarbonyl)pyrrolidine-2-carboxylic acid
A416299-1G
A416299-5G
A416299-100MG
A416299-10G
A416299-250MG
103827-314EA
216
USD
103827-314
103838-360
76841-944
76841-946
77143-096
77143-098
77143-100
N-Boc-trans-4-(fmoc-amino)-L-proline 95%
N-Boc-trans-4-(fmoc-amino)-L-proline
Precaución: for Research Use Only
Formula:
C₂₅H₂₈N₂O₆ Melting Pt: 199 °C Flash Pt: 347 °C Storage Temperature: Ambient |
MDL Number:
MFCD00673783 CAS Number: 176486-63-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...