(2S,4R)-4-Aminopyrrolidine-2-carboxylic acid, 4-BOC, N1-FMOC protected 97%

Supplier: Ambeed
Warning

Synonyms: (2S,4R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-((tert-butoxycarbonyl)amino)pyrrolidine-2-carboxylic acid, Fmoc-trans-4-N-Boc-Pro-OH

A354109-100MG A354109-250MG A354109-1G A354109-5G
103826-116EA 593 USD
103826-116 103837-120 77167-742 77167-744 77306-654 77306-656
(2S,4R)-4-Aminopyrrolidine-2-carboxylic acid, 4-BOC, N1-FMOC protected 97%
(2S,4R)-4-Aminopyrrolidine-2-carboxylic acid, 4-BOC, N1-FMOC protected

Precaución: for Research Use Only

Formula: C₂₅H₂₈N₂O₆
Boiling Pt: 650.5±55.0 °C (760 mm Hg)
Melting Pt: 85…87 °C
Flash Pt: 347.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01860707
CAS Number: 273222-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR