D-(-)-Diethyl tartrate 98%

Supplier: Ambeed
Warning

Synonyms: (2S,3S)-Diethyl 2,3-dihydroxysuccinate, (S,S)-(-)-Diethyl tartrate, (2S,3S)-(-)-Diethyl tartrate, (-)-Diethyl D-tartrate, (S,S)-(-)-Tartaric acid diethyl ester, D(-)-Tartaric acid diethyl ester

A192576-100G A192576-10G A192576-500G A192576-1000G A192576-25G
103809-274EA 151 USD
103809-274 103847-068 76800-022 76800-024 76800-026 77084-940 77302-300
D-(-)-Diethyl tartrate 98%
D-(-)-Diethyl tartrate

Precaución: for Research Use Only

Formula: C₈H₁₄O₆
Boiling Pt: 280 °C (760 mm Hg)
Melting Pt: 17 °C
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00064451
CAS Number: 13811-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR