(2S,3S)-3-Hydroxypyrrolidine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A121116-100MG A121116-10G A121116-250MG A121116-5G A121116-1G
103800-030EA 230 USD
103800-030 76766-488 76766-490 76766-492 76766-494 77312-148
(2S,3S)-3-Hydroxypyrrolidine-2-carboxylic acid 97%
(2S,3S)-3-Hydroxypyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₉NO₃
MW: 131.13 g/mol
Boiling Pt: 355.2 °C (760 mm Hg)
Melting Pt: 235 °C
Flash Pt: 168.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00216471
CAS Number: 4298-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR