(2S,3S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxybutanoic acid 95%

Supplier: Ambeed
Warning

A280576-5G A280576-100MG A280576-250MG A280576-1G
103813-512EA 24 USD
103813-512 103853-458 76837-148 76875-300 76875-302 77093-390
(2S,3S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxybutanoic acid 95%
(2S,3S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxybutanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₇NO₅
MW: 219.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23134619
CAS Number: 23082-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR