Boc-N-methyl-L-isoleucine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2S,3S)-2-((tert-Butoxycarbonyl)(methyl)amino)-3-methylpentanoic acid, Boc-N-Me-Ile-OH
A128264-10G
A128264-1G
A128264-5G
A128264-100G
103801-098EA
23
USD
103801-098
77094-860
77313-400
77313-402
77685-113
Boc-N-methyl-L-isoleucine 95%
Boc-N-methyl-L-isoleucine
Precaución: for Research Use Only
Formula:
C₁₂H₂₃NO₄ MW: 245.319 g/mol Boiling Pt: 338.2 °C (760 mm Hg) Flash Pt: 158.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00066105 CAS Number: 52498-32-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...