1,2,3-Tri-O-acetyl-5-deoxy-β-D-ribofuranose 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2S,3R,4R,5R)-5-Methyltetrahydrofuran-2,3,4-triyl triacetate
A105837-100G
A105837-10G
A105837-1G
A105837-25G
A105837-500G
A105837-5G
103822-698EA
6
USD
103822-698
77107-600
77107-602
77107-604
77107-606
77107-608
77283-404
1,2,3-Tri-O-acetyl-5-deoxy-β-D-ribofuranose 97%
1,2,3-Tri-O-acetyl-5-deoxy-beta-D-ribofuranose
Precaución: for Research Use Only
Formula:
C₁₁H₁₆O₇ MW: 260.24 g/mol Boiling Pt: 315.3 °C (760 mm Hg) Melting Pt: 63…64 °C Flash Pt: 136 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08458459 CAS Number: 62211-93-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...