1,2,3,5-Tetra-O-benzoyl-2-C-methyl-β-D-ribofuranose 98%

Supplier: Ambeed
Warning

Synonyms: (2S,3R,4R,5R)-2,4-bis(benzoyloxy)-5-[(benzoyloxy)methyl]-3-methyloxolan-3-yl benzoate, (2S,3R,4R,5R)-5-((Benzoyloxy)methyl)-3-methyltetrahydrofuran-2,3,4-triyl tribenzoate

A158762-10G A158762-25G A158762-1G A158762-5G
103805-168EA 19 USD
103805-168 103843-382 77163-112 77163-114 77300-364 77300-366
1,2,3,5-Tetra-O-benzoyl-2-C-methyl-β-D-ribofuranose 98%
1,2,3,5-Tetra-O-benzoyl-2-C-methyl-beta-D-ribofuranose

Precaución: Sealed in dry, store in freezer, under –20 °C.

Formula: C₃₄H₂₈O₉
MW: 580.589 g/mol
Melting Pt: 154…156 °C
Storage Temperature: Freezer
MDL Number: MFCD07369658
CAS Number: 15397-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR