(5S,6R)-(+)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone 95%

Supplier: Ambeed
Warning

Synonyms: Benzyl (2S,3R)-(+)-6-oxo-2,3-diphenyl-4-morpholinecarboxylate, (2S,3R)-benzyl 6-oxo-2,3- diphenylmorpholine-4-carboxylate, (2S,3R)-Benzyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate

A180131-100MG A180131-10G A180131-250MG A180131-1G A180131-5G
103807-514EA 36 USD
103807-514 103845-868 76998-638 76998-640 76998-642 77083-354 77083-356
(5S,6R)-(+)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone 95%
(5S,6R)-(+)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone

Precaución: for Research Use Only

Formula: C₂₄H₂₁NO₄
MW: 387.435 g/mol
Melting Pt: 205…207 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00074956
CAS Number: 105228-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR