(2S,3aS,6aS)-Benzyl octahydrocyclopenta[b]pyrrole-2-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Benzyl (S,S,S)-2-Azabicyclo[3.3.0]octane-3-carboxylate HCl, Benzyl (S,S,S)-2-Azabicyclo[3.3.0]octane-3-carboxylate hydrochloride, (S,S,S)-2-Azabicyclo[3.3.0]octane-3-carboxylic acid Benzyl ester hydrochloride

A132463-1G A132463-100G A132463-25G
103801-742EA 20 USD
103801-742 77298-560 77666-904 77871-624
(2S,3aS,6aS)-Benzyl octahydrocyclopenta[b]pyrrole-2-carboxylate hydrochloride 97%
(2S,3aS,6aS)-Benzyl octahydrocyclopenta[b]pyrrole-2-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₂₀ClNO₂
MW: 281.7800 g/mol
Melting Pt: 176…180 °C
Storage Temperature: Ambient
MDL Number: MFCD04974076
CAS Number: 87269-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR