(2R,5S)-2,5-Dimethylpiperazine, N1-BOC protected 95%

Supplier: Ambeed
Warning

A154608-100G A154608-10G A154608-250MG A154608-1G A154608-25G A154608-5G
103804-662EA 10 USD
103804-662 103842-852 76783-536 76783-538 76783-540 77318-584 77318-586 77318-588
(2R,5S)-2,5-Dimethylpiperazine, N1-BOC protected 95%
(2R,5S)-2,5-Dimethylpiperazine, N1-BOC protected

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
Boiling Pt: 280 °C (760 mm Hg)
Flash Pt: 123 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06797728
CAS Number: 309915-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR