tert-Butyl (2R,5R)-2,5-dimethylpiperazine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: (2R,5R)-tert-Butyl 2,5-dimethylpiperazine-1-carboxylate

A259658-100MG A259658-250MG A259658-1G A259658-5G
103812-456EA 179 USD
103812-456 103852-338 76874-828 76874-830 77091-912 77091-914
tert-Butyl (2R,5R)-2,5-dimethylpiperazine-1-carboxylate 95%
tert-Butyl (2R,5R)-2,5-dimethylpiperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.305 g/mol
Boiling Pt: 280 °C (760 mm Hg)
Flash Pt: 123 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08686669
CAS Number: 1240586-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR